Is Amberlite XAD-16 (Cat. No. L19565.22, L19565.36) a styrene-based copolymer and what is the monomer structure?
Amberlite XAD-16 (Cat. No. L19565.22, L19565.36) is a divinylbenzene-crosslinked copolymer based on styrene.
Search
View additional product information for Amberlite XAD-16 - FAQs (L19565.36, L19565.22)
1 product FAQs found