Amberlite™ IRA-900(Cl), ion exchange resin
Amberlite™ IRA-900(Cl), ion exchange resin
Amberlite™ IRA-900(Cl), ion exchange resin
Thermo Scientific Chemicals

Amberlite™ IRA-900(Cl), ion exchange resin

CAS: 9050-97-9 | C4H12ClN | 109.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number L19567.0B
also known as L19567-0B
Price (USD)
99.65
Online exclusive
111.00
Save 11.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
99.65
Online exclusive
111.00
Save 11.35
Each
Chemical Identifiers
CAS9050-97-9
MDL NumberMFCD00132712
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
FormBeads
Harmonic mean size0.650-0.820 mm
Moisture holding capacity58.0 to 64.0% (Cl⁻ form)
Particle Size≤0.5% (<0.300 mm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only