(R)-(-)-2-Chloromandelic acid, ChiPros 99+%, ee 99+%
(R)-(-)-2-Chloromandelic acid, ChiPros 99+%, ee 99+%
(R)-(-)-2-Chloromandelic acid, ChiPros 99+%, ee 99+%
Thermo Scientific Chemicals

(R)-(-)-2-Chloromandelic acid, ChiPros 99+%, ee 99+%

CAS: 52950-18-2 | C8H7ClO3 | 186.591 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L19586.03
also known as L19586-03
Price (USD)
29.65
Special offer
Online exclusive
Ends: 30-Jun-2026
34.80
Save 5.15 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
29.65
Special offer
Online exclusive
Ends: 30-Jun-2026
34.80
Save 5.15 (15%)
Each
Chemical Identifiers
CAS52950-18-2
IUPAC Name2-(2-chlorophenyl)-2-hydroxyacetic acid
Molecular FormulaC8H7ClO3
InChI KeyRWOLDZZTBNYTMS-UHFFFAOYNA-N
SMILESOC(C(O)=O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Formcrystalline powder
Assay (Aqueous acid-base Titration)>99.0%
Enantiomeric excess>99.0%
Optical Rotation-123 ? 2? (c=3 in water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only