3-Bromobenzaldehyde ethylene acetal, 98+%
3-Bromobenzaldehyde ethylene acetal, 98+%
3-Bromobenzaldehyde ethylene acetal, 98+%
Thermo Scientific Chemicals

3-Bromobenzaldehyde ethylene acetal, 98+%

CAS: 17789-14-9 | C9H9BrO2 | 229.073 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19611.06
also known as L19611-06
Price (USD)
36.65
Online Exclusive
40.70
Save 4.05 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
36.65
Online Exclusive
40.70
Save 4.05 (10%)
Each
Chemical Identifiers
CAS17789-14-9
IUPAC Name2-(3-bromophenyl)-1,3-dioxolane
Molecular FormulaC9H9BrO2
InChI KeyVYPYKCPWNPPBBX-UHFFFAOYSA-N
SMILESBrC1=CC=CC(=C1)C1OCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5605-1.5645 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only