5-Bromo-2-cyanopyridine, 95%, Thermo Scientific Chemicals
5-Bromo-2-cyanopyridine, 95%, Thermo Scientific Chemicals
5-Bromo-2-cyanopyridine, 95%, Thermo Scientific Chemicals
5-Bromo-2-cyanopyridine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-cyanopyridine, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L19621.06
also known as L19621-06
5 g
Catalog number L19621.06
also known as L19621-06
Price (USD)
49.65
Online Exclusive
55.50
Save 5.85 (11%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
55.50
Save 5.85 (11%)
Each
Add to cart
Chemical Identifiers
CAS97483-77-7
IUPAC Name5-bromopyridine-2-carbonitrile
Molecular FormulaC6H3BrN2
InChI KeyDMSHUVBQFSNBBL-UHFFFAOYSA-N
SMILESBrC1=CC=C(N=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥94.0%
Identification (FTIR)Conforms
It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol

Notes
Store in cool place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jinhua J. Song.; Nathan K. Yee.; Zhulin Tan.; Jinghua Xu.; Suresh R. Kapadia.; Chris H. Senanayake. Synthesis of 5-Bromopyridyl-2-magnesium Chloride and Its Application in the Synthesis of Functionalized Pyridines.Org. Lett. 2004, 6 (26),4905-4907 .
  2. Danuta Branowska.; Abdelbasset A. Farahat.; Arvind Kumar.; Tanja Wenzler.; Reto Brun.; Yang Liu.; W. David Wilson.; David W. Boykina. Synthesis and antiprotozoal activity of 2,5-bis[amidinoaryl]thiazoles.Bioorg. Med. Chem. 2010, 18 (10),3551-3558 .