2-Chloro-4-iodopyridine, 98%
2-Chloro-4-iodopyridine, 98%
2-Chloro-4-iodopyridine, 98%
2-Chloro-4-iodopyridine, 98%
Thermo Scientific Chemicals

2-Chloro-4-iodopyridine, 98%

CAS: 153034-86-7 | C5H3ClIN | 239.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19623.03
also known as L19623-03
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.20
Save 6.55 (16%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.20
Save 6.55 (16%)
Each
Add to cart
Chemical Identifiers
CAS153034-86-7
IUPAC Name2-chloro-4-iodopyridine
Molecular FormulaC5H3ClIN
InChI KeyKJKIPRQNFDUULB-UHFFFAOYSA-N
SMILESClC1=CC(I)=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.5%
Melting Point (clear melt)40.0-47.0?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only