Isoquinoline-5-carboxaldehyde, 97%
Isoquinoline-5-carboxaldehyde, 97%
Isoquinoline-5-carboxaldehyde, 97%
Thermo Scientific Chemicals

Isoquinoline-5-carboxaldehyde, 97%

CAS: 80278-67-7 | C10H7NO | 157.172 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19636.03
also known as L19636-03
Price (USD)
78.65
Special offer
Online exclusive
Ends: 28-Dec-2026
92.30
Save 13.65
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
78.65
Special offer
Online exclusive
Ends: 28-Dec-2026
92.30
Save 13.65
Each
Chemical Identifiers
CAS80278-67-7
IUPAC Nameisoquinoline-5-carbaldehyde
Molecular FormulaC10H7NO
InChI KeyILRSABOCKMOFGW-UHFFFAOYSA-N
SMILESO=CC1=C2C=CN=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Yellow to brown
FormPowder
Free acid (titration)≤3%
Melting Point (clear melt)113.0-122.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only