Quinoline-8-boronic acid, tech. 90%
Quinoline-8-boronic acid, tech. 90%
Quinoline-8-boronic acid, tech. 90%
Quinoline-8-boronic acid, tech. 90%
Thermo Scientific Chemicals

Quinoline-8-boronic acid, tech. 90%

CAS: 86-58-8 | C9H8BNO2 | 172.978 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19640.03
also known as L19640-03
Price (USD)
129.65
Online Exclusive
144.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
129.65
Online Exclusive
144.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS86-58-8
IUPAC Name(quinolin-8-yl)boronic acid
Molecular FormulaC9H8BNO2
InChI KeyKXJJSKYICDAICD-UHFFFAOYSA-N
SMILESOB(O)C1=C2N=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to yellow to brown
FormPowder
Assay (Aqueous acid-base Titration)≥88.0%
Assay (HPLC)≥88.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only