(E)-alpha,beta-Styrenediboronic acid bis(pinacol) ester, 98%, Thermo Scientific Chemicals
(E)-alpha,beta-Styrenediboronic acid bis(pinacol) ester, 98%, Thermo Scientific Chemicals
(E)-alpha,beta-Styrenediboronic acid bis(pinacol) ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(E)-alpha,beta-Styrenediboronic acid bis(pinacol) ester, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L19651.03
also known as L19651-03
1 g
Catalog number L19651.03
also known as L19651-03
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS173603-23-1
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)69.0-75.0?C
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only