This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L19679.03
also known as L19679-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS31886-58-5
Chemical Name or Material(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine
Density1.222
Flash Point>110°C (230°F)
Formula Weight257.16
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.