4-Methyl-beta-styrylboronic acid pinacol ester, 98%
4-Methyl-beta-styrylboronic acid pinacol ester, 98%
4-Methyl-beta-styrylboronic acid pinacol ester, 98%
Thermo Scientific Chemicals

4-Methyl-beta-styrylboronic acid pinacol ester, 98%

CAS: 149777-84-4 | C15H21BO2 | 244.141 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19698.MD
also known as L19698-MD
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Specifications
Beilstein7204616
CAS149777-84-4
Chemical Name or Material4-Methyl-beta-styrylboronic acid pinacol ester
Formula Weight244.14
IUPAC Name4,4,5,5-tetramethyl-2-[(E)-2-(4-methylphenyl)ethenyl]-1,3,2-dioxaborolane
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only