4-Cyano-2-fluorobenzoic acid, 98%
4-Cyano-2-fluorobenzoic acid, 98%
4-Cyano-2-fluorobenzoic acid, 98%
Thermo Scientific Chemicals

4-Cyano-2-fluorobenzoic acid, 98%

CAS: 164149-28-4 | C8H4FNO2 | 165.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19722.06
also known as L19722-06
Price (USD)
95.65
Online Exclusive
106.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
95.65
Online Exclusive
106.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS164149-28-4
IUPAC Name4-cyano-2-fluorobenzoic acid
Molecular FormulaC8H4FNO2
InChI KeyKEJMSTJTAWACNI-UHFFFAOYSA-N
SMILESOC(=O)C1=C(F)C=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)214.0-220.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
R.J. Mandle.; E.J. Davis.; C.-C.A. Voll.; C.T. Archbold.; J.W. Goodby.; S.J. Cowling. The relationship between molecular structure and the incidence of the NTB phase. Liquid Crystals. 2015, 42 (5-6), 688-703.