5-Bromo-2-nitropyridine, 98+%, Thermo Scientific Chemicals
5-Bromo-2-nitropyridine, 98+%, Thermo Scientific Chemicals
5-Bromo-2-nitropyridine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-nitropyridine, 98+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L19748.06
also known as L19748-06
5 g
Catalog number L19748.06
also known as L19748-06
Price (USD)
93.65
Online exclusive
104.00
Save 10.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
93.65
Online exclusive
104.00
Save 10.35
Each
Chemical Identifiers
CAS39856-50-3
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)146.0-152.0?C
Appearance (Color)Pale yellow to yellow
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only