1-Ethyl-3-methylimidazolium tetrafluoroborate, 98+% (dry wt.), may cont. up to 3% water
1-Ethyl-3-methylimidazolium tetrafluoroborate, 98+% (dry wt.), may cont. up to 3% water
1-Ethyl-3-methylimidazolium tetrafluoroborate, 98+% (dry wt.), may cont. up to 3% water
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium tetrafluoroborate, 98+% (dry wt.), may cont. up to 3% water

CAS: 143314-16-3 | C6H11BF4N2 | 197.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number L19763.18
also known as L19763-18
Price (USD)
371.65
Online Exclusive
413.00
Save 41.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
371.65
Online Exclusive
413.00
Save 41.35 (10%)
Each
Chemical Identifiers
CAS143314-16-3
IUPAC Name1-ethyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Molecular FormulaC6H11BF4N2
InChI KeyCUNYTRQQXKCRTJ-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to orange
Assay from Supplier's CofA≥98.0% (NMR)
Assay from Supplier's CofAHalides: ≤1% (IC)
Water Content (Karl Fischer Titration)<3.0%
Assay from Supplier's CofA1-Ethyl-3-methylimidazolium: ≥98.0% (IC)
View more

1-Ethyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used in the recycling of osmium in the dihydroxylation of olefins with osmium(VIII)¬ oxide. It is a stable liquid electrolyte used in the capillary electrophoretic method for resolving phenolic compounds present in grape seed extract. It also acts as a good solvent for a wide range of both organic and inorganic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used in the recycling of osmium in the dihydroxylation of olefins with osmium(VIII)¬ oxide. It is a stable liquid electrolyte used in the capillary electrophoretic method for resolving phenolic compounds present in grape seed extract. It also acts as a good solvent for a wide range of both organic and inorganic compounds.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and water.
RUO – Research Use Only

General References:

  1. Ionic liquid.
  2. Recycling of Os in the dihydroxylation of olefins with Osmium(VIII) oxide, 12103, can be accomplished by the use of this ionic liquid: Tetrahedron Lett., 43, 6849 (2002).
  3. Tooming, T.; Thomberg, T.; Kurig, H.; Jänes, A.; Lust, E. High power density supercapacitors based on the carbon dioxide activated d-glucose derived carbon electrodes and 1-ethyl-3-methylimidazolium tetrafluoroborate ionic liquid. J. power sources 2015, 280, 667-677.
  4. Thürmer, S.; Kobayashi, Y.; Ohba, T.; Kanoh, H. Pore-size dependent effects on structure and vibrations of 1-ethyl-3-methylimidazolium tetrafluoroborate in nanoporous carbon. Chem. Phys. Lett. 2015, 636, 129-133.