3-(Trifluoromethoxy)benzeneboronic acid, 98%
3-(Trifluoromethoxy)benzeneboronic acid, 98%
3-(Trifluoromethoxy)benzeneboronic acid, 98%
Thermo Scientific Chemicals

3-(Trifluoromethoxy)benzeneboronic acid, 98%

CAS: 179113-90-7 | C7H6BF3O3 | 205.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19775.03
also known as L19775-03
Price (USD)
57.65
Online exclusive
63.70
Save 6.05
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
57.65
Online exclusive
63.70
Save 6.05
Each
Chemical Identifiers
CAS179113-90-7
IUPAC Name[3-(trifluoromethoxy)phenyl]boronic acid
Molecular FormulaC7H6BF3O3
InChI KeyUWDFWVLAHRQSKK-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC(OC(F)(F)F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5%
Proton NMRConforms to structure
Appearance (Color)White
FormPowder
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only