3-Fluoropiperidine hydrochloride, 97%
3-Fluoropiperidine hydrochloride, 97%
3-Fluoropiperidine hydrochloride, 97%
Thermo Scientific Chemicals

3-Fluoropiperidine hydrochloride, 97%

CAS: 737000-77-0 | C5H11ClFN | 139.598 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19776.MD
also known as L19776-MD
Price (USD)
81.65
Special offer
Online exclusive
Ends: 30-Jun-2026
95.70
Save 14.05 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
81.65
Special offer
Online exclusive
Ends: 30-Jun-2026
95.70
Save 14.05 (15%)
Each
Chemical Identifiers
CAS737000-77-0
IUPAC Name3-fluoropiperidine hydrochloride
Molecular FormulaC5H11ClFN
InChI KeyRDJUBLSLAULIAT-UHFFFAOYNA-N
SMILESCl.FC1CCCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormPowder or lumps
Water Content (Karl Fischer Titration)≤1.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only