Methyl 2-amino-5-fluorobenzoate, 98%
Methyl 2-amino-5-fluorobenzoate, 98%
Methyl 2-amino-5-fluorobenzoate, 98%
Thermo Scientific Chemicals

Methyl 2-amino-5-fluorobenzoate, 98%

CAS: 319-24-4 | C8H8FNO2 | 169.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19778.14
also known as L19778-14
Price (USD)
189.65
Online Exclusive
210.00
Save 20.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
189.65
Online Exclusive
210.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS319-24-4
IUPAC Namemethyl 2-amino-5-fluorobenzoate
Molecular FormulaC8H8FNO2
InChI KeyPUDDYSBKCDKATP-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(N)C=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear cream to yellow to green to brown
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5560-1.5600 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only