2-Amino-6-chloropyridine, 98%
2-Amino-6-chloropyridine, 98%
2-Amino-6-chloropyridine, 98%
Thermo Scientific Chemicals

2-Amino-6-chloropyridine, 98%

CAS: 45644-21-1 | C5H5ClN2 | 128.559 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L19836.06
also known as L19836-06
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.00
Save 13.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.00
Save 13.35 (15%)
Each
Chemical Identifiers
CAS45644-21-1
IUPAC Name6-chloropyridin-2-amine
Molecular FormulaC5H5ClN2
InChI KeyOBYJTLDIQBWBHM-UHFFFAOYSA-N
SMILESNC1=CC=CC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Melting Point (clear melt)67-75?C
Appearance (Color)White to cream to brown or pale yellow to yellow or grey
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only