2,4,6-Trimethoxybenzeneboronic acid, 98%
2,4,6-Trimethoxybenzeneboronic acid, 98%
2,4,6-Trimethoxybenzeneboronic acid, 98%
2,4,6-Trimethoxybenzeneboronic acid, 98%
Thermo Scientific Chemicals

2,4,6-Trimethoxybenzeneboronic acid, 98%

CAS: 135159-25-0 | C9H13BO5 | 212.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19837.03
also known as L19837-03
Price (USD)
78.65
Special offer
Online exclusive
Ends: 30-Jun-2026
92.50
Save 13.85 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
78.65
Special offer
Online exclusive
Ends: 30-Jun-2026
92.50
Save 13.85 (15%)
Each
Add to cart
Chemical Identifiers
CAS135159-25-0
IUPAC Name(2,4,6-trimethoxyphenyl)boronic acid
Molecular FormulaC9H13BO5
InChI KeyPKLRXZVPEQJTTJ-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=C(B(O)O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
Assay (HPLC)≥97.5%
Proton NMRConforms to structure
Melting Point (clear melt)96.0-103.0?C
FormCrystals or powder or crystalline powder
2,4,6-Trimethoxybenzeneboronic acid is used as an intermediate in organic syntheses and in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxybenzeneboronic acid is used as an intermediate in organic syntheses and in Suzuki reaction.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Incompatible with oxidizing agents.
RUO – Research Use Only