2-Bromopyridine-4-carboxylic acid, 97%
2-Bromopyridine-4-carboxylic acid, 97%
2-Bromopyridine-4-carboxylic acid, 97%
Thermo Scientific Chemicals

2-Bromopyridine-4-carboxylic acid, 97%

CAS: 66572-56-3 | C6H4BrNO2 | 202.007 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19855.06
also known as L19855-06
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS66572-56-3
IUPAC Name2-bromopyridine-4-carboxylic acid
Molecular FormulaC6H4BrNO2
InChI KeyYBTKGKVQEXAYEM-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=NC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream to brown
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only