2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%
2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%
2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%

CAS: 681812-07-7 | C8H5BF6O2 | 257.926 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L19952.03
also known as L19952-03
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.70
Save 13.05 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.70
Save 13.05 (14%)
Each
Chemical Identifiers
CAS681812-07-7
IUPAC Name[2,6-bis(trifluoromethyl)phenyl]boronic acid
Molecular FormulaC8H5BF6O2
InChI KeyWAMPGNNEOZGBHR-UHFFFAOYSA-N
SMILESOB(O)C1=C(C=CC=C1C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to yellow
FormPowder
Assay (HPLC)≥96.0%
Proton NMRConforms to structure
2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only