4-(Bromomethyl)benzeneboronic acid, tech. 85%
4-(Bromomethyl)benzeneboronic acid, tech. 85%
4-(Bromomethyl)benzeneboronic acid, tech. 85%
Thermo Scientific Chemicals

4-(Bromomethyl)benzeneboronic acid, tech. 85%

CAS: 68162-47-0 | C7H8BBrO2 | 214.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19953.06
also known as L19953-06
Price (USD)
303.65
Online Exclusive
337.00
Save 33.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
303.65
Online Exclusive
337.00
Save 33.35 (10%)
Each
Chemical Identifiers
CAS68162-47-0
IUPAC Name[4-(bromomethyl)phenyl]boronic acid
Molecular FormulaC7H8BBrO2
InChI KeyPDNOURKEZJZJNZ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (HPLC)≥83.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only