4-(Bromomethyl)benzeneboronic acid pinacol ester, 95%
4-(Bromomethyl)benzeneboronic acid pinacol ester, 95%
4-(Bromomethyl)benzeneboronic acid pinacol ester, 95%
Thermo Scientific Chemicals

4-(Bromomethyl)benzeneboronic acid pinacol ester, 95%

CAS: 138500-85-3 | C13H18BBrO2 | 297.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19954.03
also known as L19954-03
Price (USD)
117.65
Online Exclusive
131.00
Save 13.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
117.65
Online Exclusive
131.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS138500-85-3
IUPAC Name2-[4-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC13H18BBrO2
InChI KeyCBUOGMOTDGNEAW-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)80-89?C
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only