2-Nitrobenzeneboronic acid pinacol ester, 98+%
2-Nitrobenzeneboronic acid pinacol ester, 98+%
2-Nitrobenzeneboronic acid pinacol ester, 98+%
Thermo Scientific Chemicals

2-Nitrobenzeneboronic acid pinacol ester, 98+%

CAS: 190788-59-1 | C12H16BNO4 | 249.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19963.03
also known as L19963-03
Price (USD)
63.65
Online Exclusive
70.50
Save 6.85 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.50
Save 6.85 (10%)
Each
Chemical Identifiers
CAS190788-59-1
IUPAC Name4,4,5,5-tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane
Molecular FormulaC12H16BNO4
InChI KeyVLJYUDGCEKORNG-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to dark brown
FormCrystalline powder
Assay (GC)>98.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only