2-(2-Fluorophenyl)ethanol, 99%
2-(2-Fluorophenyl)ethanol, 99%
2-(2-Fluorophenyl)ethanol, 99%
Thermo Scientific Chemicals

2-(2-Fluorophenyl)ethanol, 99%

CAS: 50919-06-7 | C8H9FO | 140.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19998.06
also known as L19998-06
Price (USD)
67.65
Online Exclusive
75.40
Save 7.75 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
67.65
Online Exclusive
75.40
Save 7.75 (10%)
Each
Chemical Identifiers
CAS50919-06-7
IUPAC Name2-(2-fluorophenyl)ethan-1-ol
Molecular FormulaC8H9FO
InChI KeyHNIGZVZDWCTFPR-UHFFFAOYSA-N
SMILESOCCC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5075-1.5110 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only