5-Amino-2-bromopyridine, 97%
5-Amino-2-bromopyridine, 97%
5-Amino-2-bromopyridine, 97%
Thermo Scientific Chemicals

5-Amino-2-bromopyridine, 97%

CAS: 13534-97-9 | C5H5BrN2 | 173.013 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L20008.14
also known as L20008-14
Price (USD)
431.65
Special offer
Online exclusive
Ends: 30-Jun-2026
508.00
Save 76.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
431.65
Special offer
Online exclusive
Ends: 30-Jun-2026
508.00
Save 76.35 (15%)
Each
Chemical Identifiers
CAS13534-97-9
IUPAC Name6-bromopyridin-3-amine
Molecular FormulaC5H5BrN2
InChI KeyXTHKRYHULUJQHN-UHFFFAOYSA-N
SMILESNC1=CC=C(Br)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)73.0-82.0?C
Appearance (Color)Yellow to green to brown to gray
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only