(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate, 97%
(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate, 97%
(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate, 97%
Thermo Scientific Chemicals

(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate, 97%

CAS: 737000-89-4 | C14H17NOS | 247.356 g/mol
Have Questions?
Catalog NumberQuantity
L20060.03
also known as L20060-03
1 g
Catalog number L20060.03
also known as L20060-03
Price (USD)
92.65
Special offer
Online exclusive
Ends: 30-Jun-2026
109.00
Save 16.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 30-Jun-2026
109.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS737000-89-4
IUPAC Name{[(2-isothiocyanatocyclohexyl)oxy]methyl}benzene
Molecular FormulaC14H17NOS
InChI KeyDNRPSBSMXLYTQF-UHFFFAOYNA-N
SMILESS=C=NC1CCCCC1OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5655-1.5705 @ 20?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only