6-Bromopyridine-3-boronic acid, 95%
6-Bromopyridine-3-boronic acid, 95%
6-Bromopyridine-3-boronic acid, 95%
Thermo Scientific Chemicals

6-Bromopyridine-3-boronic acid, 95%

CAS: 223463-14-7 | C5H5BBrNO2 | 201.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L20085.06
also known as L20085-06
Price (USD)
549.65
Online Exclusive
610.00
Save 60.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
549.65
Online Exclusive
610.00
Save 60.35 (10%)
Each
Chemical Identifiers
CAS223463-14-7
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms to structure
FormPowder
Assay (HPLC)≥94.0%
Appearance (Color)Cream
Assay (Aqueous acid-base Titration)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only