2-Methoxypyridine-5-boronic acid, 95%
2-Methoxypyridine-5-boronic acid, 95%
2-Methoxypyridine-5-boronic acid, 95%
2-Methoxypyridine-5-boronic acid, 95%
Thermo Scientific Chemicals

2-Methoxypyridine-5-boronic acid, 95%

CAS: 163105-89-3 | C6H8BNO3 | 152.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20087.06
also known as L20087-06
Price (USD)
183.65
Special offer
Online exclusive
Ends: 30-Jun-2026
216.00
Save 32.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
183.65
Special offer
Online exclusive
Ends: 30-Jun-2026
216.00
Save 32.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS163105-89-3
IUPAC Name(6-methoxypyridin-3-yl)boronic acid
Molecular FormulaC6H8BNO3
InChI KeyDHADXDMPEUWEAS-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=N1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥94.0%
FormPowder
Proton NMRConforms to structure
2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
RUO – Research Use Only
Gayatri Sarmah.; Utpal Bora. Simple aminobenzoic acid promoted palladium catalyzed room temperature Suzuki-Miyaura cross-coupling reaction in aqueous media. Tetrahedron Letters. 2015, 56, (22), 2906-2909.