2-Methoxypyridine-3-boronic acid, 98%
2-Methoxypyridine-3-boronic acid, 98%
2-Methoxypyridine-3-boronic acid, 98%
2-Methoxypyridine-3-boronic acid, 98%
Thermo Scientific Chemicals

2-Methoxypyridine-3-boronic acid, 98%

CAS: 163105-90-6 | C6H8BNO3 | 152.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20094.03
also known as L20094-03
Price (USD)
73.65
Special offer
Online exclusive
Ends: 30-Jun-2026
86.60
Save 12.95 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
Ends: 30-Jun-2026
86.60
Save 12.95 (15%)
Each
Add to cart
Chemical Identifiers
CAS163105-90-6
IUPAC Name(2-methoxypyridin-3-yl)boronic acid
Molecular FormulaC6H8BNO3
InChI KeyNVOLYUXUHWBCRJ-UHFFFAOYSA-N
SMILESCOC1=NC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms to structure
FormPowder
Assay (HPLC)≥97.5%
Appearance (Color)White to cream to pale brown
Assay (Aqueous acid-base Titration)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only