2-Chloro-4-(trifluoromethyl)benzeneboronic acid, 96%
2-Chloro-4-(trifluoromethyl)benzeneboronic acid, 96%
2-Chloro-4-(trifluoromethyl)benzeneboronic acid, 96%
Thermo Scientific Chemicals

2-Chloro-4-(trifluoromethyl)benzeneboronic acid, 96%

CAS: 254993-59-4 | C7H5BClF3O2 | 224.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20103.03
also known as L20103-03
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.00
Save 7.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.00
Save 7.35 (15%)
Each
Chemical Identifiers
CAS254993-59-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥95.0%
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Melting Point (clear melt)109.0-118.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only