2-Chloro-5-(trifluoromethyl)benzeneboronic acid, 96%
2-Chloro-5-(trifluoromethyl)benzeneboronic acid, 96%
2-Chloro-5-(trifluoromethyl)benzeneboronic acid, 96%
Thermo Scientific Chemicals

2-Chloro-5-(trifluoromethyl)benzeneboronic acid, 96%

CAS: 182344-18-9 | C7H5BClF3O2 | 224.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20104.03
also known as L20104-03
Price (USD)
41.65
Online Exclusive
45.60
Save 3.95 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
41.65
Online Exclusive
45.60
Save 3.95 (9%)
Each
Chemical Identifiers
CAS182344-18-9
IUPAC Name[2-chloro-5-(trifluoromethyl)phenyl]boronic acid
Molecular FormulaC7H5BClF3O2
InChI KeyYVMXEHZEYONARR-UHFFFAOYSA-N
SMILESOB(O)C1=CC(=CC=C1Cl)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥95.0%
Appearance (Color)White to cream
Assay (HPLC)≥95.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)98.0-107.0?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only