2-Isopropylbenzeneboronic acid, 97%
2-Isopropylbenzeneboronic acid, 97%
2-Isopropylbenzeneboronic acid, 97%
Thermo Scientific Chemicals

2-Isopropylbenzeneboronic acid, 97%

CAS: 89787-12-2 | C9H13BO2 | 164.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20110.06
also known as L20110-06
Price (USD)
351.65
Online Exclusive
391.00
Save 39.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
351.65
Online Exclusive
391.00
Save 39.35 (10%)
Each
Chemical Identifiers
CAS89787-12-2
IUPAC Name[2-(propan-2-yl)phenyl]boronic acid
Molecular FormulaC9H13BO2
InChI KeyKTZUVUWIBZMHMC-UHFFFAOYSA-N
SMILESCC(C)C1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥96.0%
Proton NMRConforms to structure
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only