4-Amino-2,3,5,6-tetrafluorobenzoic acid, 97%
4-Amino-2,3,5,6-tetrafluorobenzoic acid, 97%
4-Amino-2,3,5,6-tetrafluorobenzoic acid, 97%
Thermo Scientific Chemicals

4-Amino-2,3,5,6-tetrafluorobenzoic acid, 97%

CAS: 944-43-4 | C7H3F4NO2 | 209.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20128.03
also known as L20128-03
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.70
Save 7.05 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.70
Save 7.05 (14%)
Each
Chemical Identifiers
CAS944-43-4
IUPAC Name4-amino-2,3,5,6-tetrafluorobenzoic acid
Molecular FormulaC7H3F4NO2
InChI KeyWTNSXWSOTDBWOR-UHFFFAOYSA-N
SMILESNC1=C(F)C(F)=C(C(O)=O)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥96.0% (U.S. specification)
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0% (non-U.S. specification)
FormPowder
Assay (HPLC)≥96.0% (non-U.S. specification)
It is used in chemical research and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in chemical research and as pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in methanol (25 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Toshifumi Takeuchi; Yuji Minato; Masayoshi Takase; Hideyuki Shinmori. Molecularly imprinted polymers with halogen bonding-based molecular recognition sites. Tetrahedron Letters. 2005, 46(52), 9025-9027
  2. Sui Xiong Cai; Denis J. Glenn; and John F. W. Keana. Toward the Development of Radiolabeled Fluorophenyl Azide-Based Photolabeling Reagents: Synthesis and Photolysis of Iodinated 4-Azidoperfluorobenzoates and 4-Azido-3,5,6-trifluorobenzoates. J. Org. Chem. 1992, 57(4), 1299-1304