(1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide, 98%
(1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide, 98%
(1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide, 98%
Thermo Scientific Chemicals

(1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide, 98%

CAS: 132747-20-7 | C5H12Br2N2 | 259.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L20170.MD
also known as L20170-MD
Price (USD)
62.65
Special offer
Online exclusive
Ends: 30-Jun-2026
73.40
Save 10.75 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 30-Jun-2026
73.40
Save 10.75 (15%)
Each
Chemical Identifiers
CAS132747-20-7
IUPAC Name2,5-diazabicyclo[2.2.1]heptane dihydrobromide
Molecular FormulaC5H12Br2N2
InChI KeyISYQWKOXKGJREA-UHFFFAOYNA-N
SMILESBr.Br.C1NC2CNC1C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown or purple
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Optical Rotation22.4 ± 0.4? (C=1 in water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only