Ethynyltri-n-butyltin, 96%
Ethynyltri-n-butyltin, 96%
Ethynyltri-n-butyltin, 96%
Thermo Scientific Chemicals

Ethynyltri-n-butyltin, 96%

CAS: 994-89-8 | C14H28Sn | 315.088 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L20178.03
also known as L20178-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS994-89-8
IUPAC Nametributyl(ethynyl)stannane
Molecular FormulaC14H28Sn
InChI KeyYEMJHNYABQHWHL-UHFFFAOYSA-N
SMILESCCCC[Sn](CCCC)(CCCC)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
FormLiquid
Appearance (Color)Clear colorless to white
Refractive Index1.4740-1.4800 @ 20?C
Used to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.

Solubility
Not miscible or difficult to mix with water.

Notes
Mositure sensitive. Store away from oxidizing agents and water/moisture.
RUO – Research Use Only
Asunciòn Barbero; Purificación Cuadrado; Ian Fleming; Ana M. González; Francisco J. Pulido; and Rosa Rubio. Stannyl-cupration of acetylenes and the reaction of the intermediate cuprates with electrophiles as a synthesis of substituted vinylstannanes. J. Chem. Soc., Perkin Trans. 1. 1993,1657-1662