Methallyltri-n-butyltin, 98%
Methallyltri-n-butyltin, 98%
Methallyltri-n-butyltin, 98%
Methallyltri-n-butyltin, 98%
Thermo Scientific Chemicals

Methallyltri-n-butyltin, 98%

CAS: 67883-62-9 | C16H34Sn | 345.158 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L20186.06
also known as L20186-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS67883-62-9
IUPAC Nametributyl(2-methylprop-2-en-1-yl)stannane
Molecular FormulaC16H34Sn
InChI KeyCYIUFVYZGOCGEV-UHFFFAOYSA-N
SMILESCCCC[Sn](CCCC)(CCCC)CC(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Refractive Index1.4845-1.4885 @ 20?C
Used as a methallylating reagent. Also used as a sole curative. It is used as a primary and secondary intermediates in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a methallylating reagent. Also used as a sole curative. It is used as a primary and secondary intermediates in pharmaceuticals.

Solubility
Fully miscible with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Mukund K. Gurjar; Somu V. Ravindranadh; Kuppusamy Sankar; Sukhen Karmakar; Joseph Cheriana; and Mukund S. Chorghade. Synthesis of spirocycles via ring closing metathesis of heterocycles carrying gem-diallyl substituents obtained via ring opening of (halomethyl)cyclopropanes with allyltributyltin. Org. Biomol. Chem.2003,(8), 1366-1373
  2. Roderick W. Bates; Attapol Pinsa; Ximin Kan. Synthesis of the Northern hemisphere of the briaranes. Tetrahedron.2003,66(33), 6340-6348