4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store under dry inert gas. Incompatible with oxidizing agents and air.
RUO – Research Use Only
Phillips AP. Cinchoninaldehyde and Aliphatic Amines—Preparation of Some Lepidylamines. J. Am. Chem. Soc.1947,69(4), 865-866