Tri-n-butyl(3-methyl-2-butenyl)tin, 95%
Tri-n-butyl(3-methyl-2-butenyl)tin, 95%
Tri-n-butyl(3-methyl-2-butenyl)tin, 95%
Thermo Scientific Chemicals

Tri-n-butyl(3-methyl-2-butenyl)tin, 95%

CAS: 53911-92-5 | C17H36Sn | 359.185 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L20214.03
also known as L20214-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS53911-92-5
IUPAC Nametributyl(3-methylbut-2-en-1-yl)stannane
Molecular FormulaC17H36Sn
InChI KeyXEFRYQPTIWSVGI-UHFFFAOYSA-N
SMILESCCCC[Sn](CCCC)(CCCC)CC=C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Refractive Index1.4855-1.4915 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Tri-n-butyl(3-methyl-2-butenyl)tin is used in prenylation of ethyl propiolate. It is also involved in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyl(3-methyl-2-butenyl)tin is used in prenylation of ethyl propiolate. It is also involved in chemical research.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Fuchs, J. R.; Funk, R. L. Indol-2-one intermediates: mechanistic evidence and synthetic utility. Total syntheses of (±)-flustramines A and C. Org. Lett. 2005, 7 (4), 677-680.
  2. Vanderhoff, P. A.; Lalancette, R. A.; Thompson, H. W. An efficient screen for determining solid-state hydrogen-bonding patterns in carboxylic acids. J. Org. Chem. 1990, 55 (6), 1696-1698.