(Triethylsilyl)acetylene, 97%
(Triethylsilyl)acetylene, 97%
(Triethylsilyl)acetylene, 97%
Thermo Scientific Chemicals

(Triethylsilyl)acetylene, 97%

CAS: 1777-03-3 | C8H16Si | 140.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20217.06
also known as L20217-06
Price (USD)
95.65
Online Exclusive
106.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
95.65
Online Exclusive
106.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS1777-03-3
IUPAC Nametriethyl(ethynyl)silane
Molecular FormulaC8H16Si
InChI KeyFWSPXZXVNVQHIF-UHFFFAOYSA-N
SMILESCC[Si](CC)(CC)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4290-1.4340 @ 20?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only