(S)-(+)-2-Octyl isocyanate, 95%
(S)-(+)-2-Octyl isocyanate, 95%
(S)-(+)-2-Octyl isocyanate, 95%
Thermo Scientific Chemicals

(S)-(+)-2-Octyl isocyanate, 95%

CAS: 745783-87-3 | C9H17NO | 155.241 g/mol
Have Questions?
Catalog NumberQuantity
L20245.03
also known as L20245-03
1 g
Catalog number L20245.03
also known as L20245-03
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS745783-87-3
IUPAC Name2-isocyanatooctane
Molecular FormulaC9H17NO
InChI KeyFTWPDIQXUDUDPL-UHFFFAOYNA-N
SMILESCCCCCCC(C)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation+37? ± 1.0? (neat)
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
Refractive Index1.4235-1.4295 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only