Cerium(IV) sulfate, anhydrous, 97%
Cerium(IV) sulfate, anhydrous, 97%
Cerium(IV) sulfate, anhydrous, 97%
Thermo Scientific Chemicals

Cerium(IV) sulfate, anhydrous, 97%

CAS: 13590-82-4 | CeO8S2 | 332.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number L20254.36
also known as L20254-36
Price (USD)
616.65
Online Exclusive
685.00
Save 68.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
616.65
Online Exclusive
685.00
Save 68.35 (10%)
Each
Chemical Identifiers
CAS13590-82-4
IUPAC Nameλ⁴-cerium(4+) disulfate
Molecular FormulaCeO8S2
InChI KeyVZDYWEUILIUIDF-UHFFFAOYSA-J
SMILES[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥96.0% (REO)
FormPowder
Impurity contentReport Metals Analysis
Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions' reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions′ reactions.

Solubility
Soluble in water and dilute sulfuric acid.

Notes
Store in cool place. Hygroscopic and moisture sensitive. Avoid moisture. Incompatible with strong reducing agents.
RUO – Research Use Only

General References:

  1. Khan, M. M. A.; Naushad, M. Synthesis, surface characterization and electrochemical properties of PVC-based cerium(IV) sulphate ion exchange composite membrane. Ionics 2015, 21 (4), 1057-1062.
  2. Asutay, O.; Hamarat, N.; Uludag, N.; Coskun, N. Selective oxidative deoximation with anhydrous Ce(IV) sulfate. Tetrahedron Lett. 2015, 56 (25), 3902-3904.