1,3-Dichloro-2-butene, cis + trans, 98%, stab. with BHT
1,3-Dichloro-2-butene, cis + trans, 98%, stab. with BHT
1,3-Dichloro-2-butene, cis + trans, 98%, stab. with BHT
Thermo Scientific Chemicals

1,3-Dichloro-2-butene, cis + trans, 98%, stab. with BHT

CAS: 926-57-8 | C4H6Cl2 | 124.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L20334.14
also known as L20334-14
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS926-57-8
IUPAC Name(2E)-1,3-dichlorobut-2-ene
Molecular FormulaC4H6Cl2
InChI KeyWLIADPFXSACYLS-DUXPYHPUSA-N
SMILESC\C(Cl)=C/CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, yellow
Formliquid
Assay (GC)>97.5% (cis+trans)
Refractive Index1.4710-1.4750 @ 20*C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only