6-Fluoropyridine-3-boronic acid, 95%
6-Fluoropyridine-3-boronic acid, 95%
6-Fluoropyridine-3-boronic acid, 95%
Thermo Scientific Chemicals

6-Fluoropyridine-3-boronic acid, 95%

CAS: 351019-18-6 | C5H5BFNO2 | 140.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L20387.03
also known as L20387-03
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
133.00
Save 19.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
133.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS351019-18-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥94.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only