6-Chloropyridine-3-boronic acid, 96%
6-Chloropyridine-3-boronic acid, 96%
6-Chloropyridine-3-boronic acid, 96%
6-Chloropyridine-3-boronic acid, 96%
Thermo Scientific Chemicals

6-Chloropyridine-3-boronic acid, 96%

CAS: 444120-91-6 | C5H5BClNO2 | 157.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20388.03
also known as L20388-03
Price (USD)
29.70
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
29.70
Each
Add to cart
Chemical Identifiers
CAS444120-91-6
IUPAC Name(6-chloropyridin-3-yl)boronic acid
Molecular FormulaC5H5BClNO2
InChI KeyWPAPNCXMYWRTTL-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Proton NMRConforms to structure
Appearance (Color)White to cream to pale yellow
Assay (Aqueous acid-base Titration)≥95.0%
Assay (HPLC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only