2,6-Dimethoxypyridine-3-boronic acid, 95%
2,6-Dimethoxypyridine-3-boronic acid, 95%
2,6-Dimethoxypyridine-3-boronic acid, 95%
Thermo Scientific Chemicals

2,6-Dimethoxypyridine-3-boronic acid, 95%

CAS: 221006-70-8 | C7H10BNO4 | 182.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L20389.06
also known as L20389-06
Price (USD)
477.65
Online Exclusive
531.00
Save 53.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
477.65
Online Exclusive
531.00
Save 53.35 (10%)
Each
Chemical Identifiers
CAS221006-70-8
IUPAC Name(2,6-dimethoxypyridin-3-yl)boronic acid
Molecular FormulaC7H10BNO4
InChI KeyADGHSWFUZUADDH-UHFFFAOYSA-N
SMILESCOC1=CC=C(B(O)O)C(OC)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥94.0%
Proton NMRConforms to structure
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only