L-(-)-Prolinamide, 98%
L-(-)-Prolinamide, 98%
L-(-)-Prolinamide, 98%
Thermo Scientific Chemicals

L-(-)-Prolinamide, 98%

CAS: 7531-52-4 | C5H11N2O | 115.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20417.03
also known as L20417-03
Price (USD)
55.65
Online Exclusive
62.10
Save 6.45 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
62.10
Save 6.45 (10%)
Each
Chemical Identifiers
CAS7531-52-4
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-105? ± 3? (c=2 in Ethanol)
FormCrystals or powder or crystalline powder
Melting Point (clear melt)95-104?C
Appearance (Color)White to pale yellow
Assay (GC)≥97.5%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also it is a efficient organocatalysts for aldol reactions on water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also it is a efficient organocatalysts for aldol reactions on water.

Solubility
Soluble in water and ethanol (50 mg/ml).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Gongkun Tang; Xianming Hu; Hans Josef Altenbach. l-Prolinamide derivatives as efficient organocatalysts for aldol reactions on water. Tetrahedron Letters. 2011, 52 (52), 7034-7037.
  2. Griet Van Zeebroeck; Beatriz Monge Bonini; Matthias Versele; Johan M Thevelein. Transport and signaling via the amino acid binding site of the yeast Gap1 amino acid transceptor. Nature Chemical Biology. 2009, 5 (1), 45-52.