Lectin PNA From Arachis hypogaea (peanut), Alexa Fluor™ 488 Conjugate
Lectin PNA is specific for terminal β-galactose. It will agglutinate human erythrocytes, but only after neuraminidase treatment.View complete list ofRead more
Have Questions?
Catalog Number
Quantity
L21409
1 mg
Catalog number L21409
Price (USD)
220.65
Online exclusive
230.00
Save 9.35
Each
Quantity:
1 mg
Price (USD)
220.65
Online exclusive
230.00
Save 9.35
Each
Lectin PNA is specific for terminal β-galactose. It will agglutinate human erythrocytes, but only after neuraminidase treatment.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor
Protein SubtypeLectins
Quantity1 mg
Shipping ConditionRoom Temperature
SourcePeanut
ConjugateAlexa Fluor 488
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Citations & References (16)
Citations & References
Abstract
Dicer inactivation leads to progressive functional and structural degeneration of the mouse retina.
Authors:Damiani D, Alexander JJ, O'Rourke JR, McManus M, Jadhav AP, Cepko CL, Hauswirth WW, Harfe BD, Strettoi E,
Journal:J Neurosci
PubMed ID:18463241
'MicroRNAs (miRNAs) are small, highly conserved molecules that have been shown to regulate the expression of genes by binding to specific target mRNAs. Dicer, an RNase III endonuclease, is essential for the production and function of mature miRNAs, and removal of Dicer has been shown to disrupt many developmental processes. ... More
Lectin mapping reveals stage-specific display of surface carbohydrates in in vitro and haemolymph-derived cells of the entomopathogenic fungus Beauveria bassiana.
Authors:Wanchoo A, Lewis MW, Keyhani NO,
Journal:Microbiology
PubMed ID:19608611
'The entomopathogenic fungus Beauveria bassiana and its insect host target represent a model system with which to examine host-pathogen interactions. Carbohydrate epitopes on the surfaces of fungal cells play diverse roles in processes that include adhesion, non-self recognition and immune invasion with respect to invertebrate hosts. B. bassiana produces a ... More
Retention of function without normal disc morphogenesis occurs in cone but not rod photoreceptors.
'It is commonly assumed that photoreceptor (PR) outer segment (OS) morphogenesis is reliant upon the presence of peripherin/rds, hereafter termed Rds. In this study, we demonstrate a differential requirement of Rds during rod and cone OS morphogenesis. In the absence of this PR-specific protein, rods do not form OSs and ... More
Mutation of the COG complex subunit gene COG7 causes a lethal congenital disorder.
Authors:Wu X, Steet RA, Bohorov O, Bakker J, Newell J, Krieger M, Spaapen L, Kornfeld S, Freeze HH
Journal:Nat Med
PubMed ID:15107842
'The congenital disorders of glycosylation (CDG) are characterized by defects in N-linked glycan biosynthesis that result from mutations in genes encoding proteins directly involved in the glycosylation pathway. Here we describe two siblings with a fatal form of CDG caused by a mutation in the gene encoding COG-7, a subunit ... More
Anti-glycosyl antibodies in lipid rafts of the enterocyte brush border: a possible host defense against pathogens.
Authors:Hansen GH, Pedersen ED, Immerdal L, Niels-Christiansen LL, Danielsen EM
Journal:Am J Physiol Gastrointest Liver Physiol
PubMed ID:16081758
The pig small intestinal brush border is a glycoprotein- and glycolipid-rich membrane that functions as a digestive/absorptive surface for dietary nutrients as well as a permeability barrier for pathogens. The present work was performed to identify carbohydrate-binding (lectinlike) proteins associated with the brush border. Chromatography on lactose-agarose was used to ... More