Lectin GS-II From Griffonia simplicifolia, Alexa Fluor™ 488 Conjugate
Lectin GS-II, isolated from the seeds of the tropical African legume Griffonia simplicifolia is a 113,000-dalton compoased of identical subunits.Read more
Have Questions?
Catalog Number
Quantity
L21415
500 μg
Catalog number L21415
Price (USD)
184.50
Special offer
Online exclusive
Ends: 31-Dec-2026
225.00
Save 40.50
Each
Quantity:
500 μg
Price (USD)
184.50
Special offer
Online exclusive
Ends: 31-Dec-2026
225.00
Save 40.50
Each
Lectin GS-II, isolated from the seeds of the tropical African legume Griffonia simplicifolia is a 113,000-dalton compoased of identical subunits. Each subunit contains a single binding site specific for terminal non-reducing a- or ß-linked N-acetyl-D-glucoasemine. Unlike many other lectins, GS-II does not react with any specific blood group. The green-fluorescent lectin GS-II Alexa Fluor™ 488 conjugate can be used in fluorescence microscopy or flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor
Protein SubtypeLectins
Quantity500 μg
Shipping ConditionRoom Temperature
SourceAfrica Shrub legume
ConjugateAlexa Fluor 488
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Citations & References (13)
Citations & References
Abstract
Lectin mapping reveals stage-specific display of surface carbohydrates in in vitro and haemolymph-derived cells of the entomopathogenic fungus Beauveria bassiana.
Authors:Wanchoo A, Lewis MW, Keyhani NO,
Journal:Microbiology
PubMed ID:19608611
'The entomopathogenic fungus Beauveria bassiana and its insect host target represent a model system with which to examine host-pathogen interactions. Carbohydrate epitopes on the surfaces of fungal cells play diverse roles in processes that include adhesion, non-self recognition and immune invasion with respect to invertebrate hosts. B. bassiana produces a ... More
The maturation of mucus-secreting gastric epithelial progenitors into digestive-enzyme secreting zymogenic cells requires Mist1.
Continuous regeneration of digestive enzyme (zymogen)-secreting chief cells is a normal aspect of stomach function that is disrupted in precancerous lesions (e.g. metaplasias, chronic atrophy). The cellular and genetic pathways that underlie zymogenic cell (ZC) differentiation are poorly understood. Here, we describe a gene expression analysis of laser capture microdissection ... More
S-nitrosoprotein formation and localization in endothelial cells.
Authors:Yang Y, Loscalzo J
Journal:Proc Natl Acad Sci U S A
PubMed ID:15618409
Protein S-nitrosation represents a recently described form of post-translational modification that is rapid and reversible. However, the analysis of protein S-nitrosation in situ has been difficult because of the absence of specific probes and the instability of cellular protein S-nitrosothiols. We developed a rapid and specific method for detecting endothelial ... More
Global mapping of glycosylation pathways in human-derived cells.
Authors:
Journal:Dev Cell
PubMed ID:33730547
βA3/A1-crystallin is a critical mediator of STAT3 signaling in optic nerve astrocytes.