Lipopolysaccharides From Escherichia coli Serotype 055:B5, Alexa Fluor™ 488 Conjugate
This bright, green-fluorescent Alexa Fluor 488 conjugate can be used to follow lipopolysaccharide binding, transport and cell-internalization processes.자세히 알아보기
Have Questions?
카탈로그 번호
수량
L23351
100 μg
카탈로그 번호 L23351
제품 가격(KRW)
381,000
온라인 행사
Ends: 30-Jun-2026
435,000
할인액 54,000 (12%)
Each
수량:
100 μg
제품 가격(KRW)
381,000
온라인 행사
Ends: 30-Jun-2026
435,000
할인액 54,000 (12%)
Each
This bright, green-fluorescent Alexa Fluor 488 conjugate can be used to follow lipopolysaccharide binding, transport and cell-internalization processes.
For Research Use Only. Not for use in diagnostic procedures.
사양
검출 방법Fluorescence
염료 유형Alexa Fluor Dyes
여기/방출495/519
형태Solid
수량100 μg
배송 조건Room Temperature
제품라인Alexa Fluor
제품 유형Lipopolysaccharide Conjugate
Unit SizeEach
구성 및 보관
Store in freezer (-5 to -30°C) and protect from light.
인용 및 참조 문헌 (16)
인용 및 참조 문헌
Abstract
Surfactant protein A directly interacts with TLR4 and MD-2 and regulates inflammatory cellular response. Importance of supratrimeric oligomerization.
'The purpose of the current study was to examine the binding of pulmonary surfactant protein A (SP-A) to TLR4 and MD-2, which are critical signaling receptors for lipopolysaccharides (LPSs). The direct binding of SP-A to the recombinant soluble form of extracellular TLR4 domain (sTLR4) and MD-2 was detected using solid-phase ... More
LPS activation of Toll-like receptor 4 signals CD11b/CD18 expression in neutrophils.
Authors:Zhou X, Gao XP, Fan J, Liu Q, Anwar KN, Frey RS, Malik AB,
Journal:Am J Physiol Lung Cell Mol Physiol
PubMed ID:15563689
We identify herein a novel signaling function of the Toll-like receptor-4 (TLR4), the lipopolysaccharide (LPS) receptor mediating the innate immune response, in inducing the expression of CD11b/CD18 integrin in polymorphonuclear leukocytes (PMNs). Studies were made in PMNs isolated from TLR4-deficient (TLR4(-/-)) and C57BL/6 [wild-type (WT)] mice. We observed increased CD11b ... More
Activation of innate immunity in the CNS triggers neurodegeneration through a Toll-like receptor 4-dependent pathway.
Innate immunity is an evolutionarily ancient system that provides organisms with immediately available defense mechanisms through recognition of pathogen-associated molecular patterns. We show that in the CNS, specific activation of innate immunity through a Toll-like receptor 4 (TLR4)-dependent pathway leads to neurodegeneration. We identify microglia as the major lipopolysaccharide (LPS)-responsive ... More
The toll-like receptor TLR4 is necessary for lipopolysaccharide-induced oligodendrocyte injury in the CNS.
The immediate or innate immune response is the first line of defense against diverse microbial pathogens and requires the expression of recently discovered toll-like receptors (TLRs). TLR4 serves as a specific receptor for lipopolysaccharide (LPS) and is localized on the surface of a subset of mammalian cells. Although innate immunity ... More
Differential type I interferon activation and susceptibility of dendritic cell populations to porcine arterivirus.
Authors:Loving CL, Brockmeier SL, Sacco RE
Journal:Immunology
PubMed ID:17116172
Dendritic cells (DCs) play a role in anti-viral immunity by providing early innate protection against viral replication and by presenting antigen to T cells for initiation of the adaptive immune response. Studies show the adaptive response to porcine reproductive and respiratory syndrome virus (PRRSV) is ineffective for complete viral elimination. ... More